Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 3 - 4 | 1425 | 88 | |
XM_021850418.1 | XP_021706110.1 | 3 - 4 | 1424 | 88 |
