Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 1 - 2 | 1145 | 67 | |
XM_021850418.1 | XP_021706110.1 | 1 - 2 | 1144 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001653754.2, XM_021850418.1 transcripts.
Set Membership: |
![]() ![]() |
