Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 9 - 10 | 2349 | 62 | |
XM_021850418.1 | XP_021706110.1 | 9 - 10 | 2348 | 62 | ||
XM_021850419.1 | XP_021706111.1 | 5 - 6 | 841 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
