Assay Details
Gene Symbol: LOC5571895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653754.2 | XP_001653804.1 | 7 - 8 | 1880 | 96 | |
XM_021850418.1 | XP_021706110.1 | 7 - 8 | 1879 | 96 | ||
XM_021850419.1 | XP_021706111.1 | 3 - 4 | 372 | 96 |
