Assay Details
Gene Symbol: LOC5579108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657255.3 | XP_001657305.2 | 2 - 3 | 290 | 68 | |
XM_021849076.1 | XP_021704768.1 | 2 - 3 | 292 | 68 |

More Information
Set Membership: |
![]() ![]() |
