Assay Details
Gene Symbol: LOC5579108 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657255.3 | XP_001657305.2 | 1 - 2 | 209 | 66 | |
XM_021849076.1 | XP_021704768.1 | 1 - 2 | 211 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001657255.3, XM_021849076.1 transcripts.
Set Membership: |
![]() ![]() |
