Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 3 - 4 | 348 | 83 | |
XM_021844764.1 | XP_021700456.1 | 3 - 4 | 460 | 83 | ||
XM_021844765.1 | XP_021700457.1 | 3 - 4 | 304 | 83 | ||
XM_021844766.1 | XP_021700458.1 | 3 - 4 | 405 | 83 | ||
XM_021844767.1 | XP_021700459.1 | 3 - 4 | 291 | 83 | ||
XM_021844768.1 | XP_021700460.1 | 2 - 3 | 176 | 83 |
