Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 9 - 10 | 2927 | 96 | |
XM_021844764.1 | XP_021700456.1 | 9 - 10 | 3039 | 96 | ||
XM_021844765.1 | XP_021700457.1 | 9 - 10 | 2883 | 96 | ||
XM_021844766.1 | XP_021700458.1 | 9 - 10 | 2984 | 96 | ||
XM_021844767.1 | XP_021700459.1 | 9 - 10 | 2870 | 96 | ||
XM_021844768.1 | XP_021700460.1 | 8 - 9 | 2755 | 96 |
