Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 8 - 9 | 2722 | 66 | |
XM_021844764.1 | XP_021700456.1 | 8 - 9 | 2834 | 66 | ||
XM_021844765.1 | XP_021700457.1 | 8 - 9 | 2678 | 66 | ||
XM_021844766.1 | XP_021700458.1 | 8 - 9 | 2779 | 66 | ||
XM_021844767.1 | XP_021700459.1 | 8 - 9 | 2665 | 66 | ||
XM_021844768.1 | XP_021700460.1 | 7 - 8 | 2550 | 66 |

More Information
Set Membership: |
![]() ![]() |
