Assay Details
Gene Symbol: LOC5569378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844763.1 | XP_021700455.1 | 6 - 7 | 1579 | 73 | |
XM_021844764.1 | XP_021700456.1 | 6 - 7 | 1691 | 73 | ||
XM_021844765.1 | XP_021700457.1 | 6 - 7 | 1535 | 73 | ||
XM_021844766.1 | XP_021700458.1 | 6 - 7 | 1636 | 73 | ||
XM_021844767.1 | XP_021700459.1 | 6 - 7 | 1522 | 73 | ||
XM_021844768.1 | XP_021700460.1 | 5 - 6 | 1407 | 73 |
