Assay Details
Gene Symbol: LOC110677245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848348.1 | XP_021704040.1 | 2 - 3 | 949 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC110677245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848348.1 | XP_021704040.1 | 2 - 3 | 949 | 95 |
Set Membership: |
![]() ![]() ![]() |