Assay Details
Gene Symbol: LOC5576036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842628.1 | XP_021698320.1 | 1 - 2 | 1056 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842628.1 transcripts.
Set Membership: |
![]() ![]() |
