Assay Details
Gene Symbol: LOC5570582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849030.1 | XP_021704722.1 | 13 - 14 | 3421 | 86 | |
XM_021849031.1 | XP_021704723.1 | 13 - 14 | 2889 | 86 | ||
XM_021849032.1 | XP_021704724.1 | 12 - 13 | 3409 | 86 | ||
XM_021849033.1 | XP_021704725.1 | 12 - 13 | 3349 | 86 |
