Assay Details
Gene Symbol: LOC5568570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849730.1 | XP_021705422.1 | 1 - 2 | 464 | 101 | |
XM_021849731.1 | XP_021705423.1 | 1 - 2 | 441 | 101 | ||
XM_021849732.1 | XP_021705424.1 | 1 - 2 | 441 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849730.1, XM_021849731.1, XM_021849732.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
