Assay Details
Gene Symbol: LOC5568570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849730.1 | XP_021705422.1 | 8 - 9 | 3421 | 107 | |
XM_021849731.1 | XP_021705423.1 | 8 - 9 | 3398 | 107 | ||
XM_021849732.1 | XP_021705424.1 | 8 - 9 | 3398 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
