Assay Details
Gene Symbol: LOC5575163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846594.1 | XP_021702286.1 | 3 - 4 | 2230 | 93 | |
XM_021846595.1 | XP_021702287.1 | 3 - 4 | 2232 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
