Assay Details
Gene Symbol: LOC5568904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841303.1 | XP_021696995.1 | 8 - 9 | 1483 | 56 | |
XM_021841304.1 | XP_021696996.1 | 8 - 9 | 1427 | 56 | ||
XM_021841305.1 | XP_021696997.1 | 8 - 9 | 1459 | 56 | ||
XM_021841307.1 | XP_021696999.1 | 8 - 9 | 1403 | 56 | ||
XM_021841308.1 | XP_021697000.1 | 8 - 9 | 1419 | 56 | ||
XM_021841309.1 | XP_021697001.1 | 8 - 9 | 1363 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
