Assay Details
Gene Symbol: LOC5568904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841303.1 | XP_021696995.1 | 5 - 6 | 991 | 72 | |
XM_021841304.1 | XP_021696996.1 | 5 - 6 | 935 | 72 | ||
XM_021841305.1 | XP_021696997.1 | 5 - 6 | 967 | 72 | ||
XM_021841307.1 | XP_021696999.1 | 5 - 6 | 911 | 72 | ||
XM_021841308.1 | XP_021697000.1 | 5 - 6 | 927 | 72 | ||
XM_021841309.1 | XP_021697001.1 | 5 - 6 | 871 | 72 |
