Assay Details
Gene Symbol: LOC110674772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839060.1 | XP_021694752.1 | 1 - 2 | 247 | 109 | |
XM_021839061.1 | XP_021694753.1 | 1 - 2 | 247 | 109 | ||
XM_021839062.1 | XP_021694754.1 | 1 - 2 | 247 | 109 | ||
XM_021839063.1 | XP_021694755.1 | 1 - 2 | 246 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021839060.1, XM_021839061.1, XM_021839062.1, XM_021839063.1 transcripts.
Set Membership: |
![]() ![]() |
