Assay Details
Gene Symbol: LOC110674772 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839060.1 | XP_021694752.1 | 10 - 11 | 4045 | 87 | |
XM_021839061.1 | XP_021694753.1 | 9 - 10 | 4039 | 87 | ||
XM_021839062.1 | XP_021694754.1 | 9 - 10 | 3790 | 87 | ||
XM_021839063.1 | XP_021694755.1 | 10 - 11 | 3294 | 87 |
