Assay Details
Gene Symbol: LOC5570514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659152.2 | XP_001659202.1 | 2 - 3 | 1069 | 79 | |
XM_021840732.1 | XP_021696424.1 | 2 - 3 | 1070 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
