Assay Details
Gene Symbol: LOC5570514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659152.2 | XP_001659202.1 | 4 - 5 | 1413 | 166 | |
XM_021840732.1 | XP_021696424.1 | 4 - 5 | 1414 | 166 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
