Assay Details
Gene Symbol: LOC5565650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649952.2 | XP_001650002.2 | 3 - 4 | 2071 | 60 | |
XM_021841214.1 | XP_021696906.1 | 3 - 4 | 2093 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
