Assay Details
Gene Symbol: LOC5565650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649952.2 | XP_001650002.2 | 1 - 2 | 501 | 57 | |
XM_021841214.1 | XP_021696906.1 | 1 - 2 | 526 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
