Assay Details
Gene Symbol: LOC5575216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850012.1 | XP_021705704.1 | 12 - 13 | 3723 | 63 | |
XM_021850013.1 | XP_021705705.1 | 12 - 13 | 3702 | 63 | ||
GenBank mRNA | JN030894.1 | - | 9 - 10 | 1782 | 63 | |
KC439534.1 | - | 9 - 10 | 2227 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
