Assay Details
Gene Symbol: LOC5575216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850012.1 | XP_021705704.1 | 8 - 9 | 2995 | 76 | |
XM_021850013.1 | XP_021705705.1 | 8 - 9 | 2995 | 76 | ||
GenBank mRNA | JN030894.1 | - | 5 - 6 | 1054 | 76 | |
KC439534.1 | - | 5 - 6 | 1499 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
