Assay Details
Gene Symbol: LOC5571516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837280.1 | XP_021692972.1 | 7 - 8 | 1039 | 94 | |
XM_021837281.1 | XP_021692973.1 | 7 - 8 | 973 | 94 | ||
XM_021837283.1 | XP_021692975.1 | 7 - 8 | 942 | 94 | ||
XM_021837284.1 | XP_021692976.1 | 6 - 7 | 1013 | 94 | ||
XM_021837285.1 | XP_021692977.1 | 6 - 7 | 993 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
