Assay Details
Gene Symbol: LOC5571516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837280.1 | XP_021692972.1 | 6 - 7 | 849 | 96 | |
XM_021837281.1 | XP_021692973.1 | 6 - 7 | 783 | 96 | ||
XM_021837283.1 | XP_021692975.1 | 6 - 7 | 752 | 96 | ||
XM_021837284.1 | XP_021692976.1 | 5 - 6 | 823 | 96 | ||
XM_021837285.1 | XP_021692977.1 | 5 - 6 | 803 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021837280.1, XM_021837281.1, XM_021837283.1, XM_021837284.1, XM_021837285.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
