Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 4 - 5 | 1723 | 91 | |
XM_021843406.1 | XP_021699098.1 | 5 - 6 | 1562 | 91 | ||
XM_021843407.1 | XP_021699099.1 | 4 - 5 | 1651 | 91 | ||
XM_021843408.1 | XP_021699100.1 | 3 - 4 | 992 | 91 |
