Assay Details
Gene Symbol: LOC5577461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663337.2 | XP_001663387.2 | 14 - 15 | 4290 | 93 | |
XM_021843406.1 | XP_021699098.1 | 15 - 16 | 4129 | 93 | ||
XM_021843407.1 | XP_021699099.1 | 14 - 15 | 4218 | 93 | ||
XM_021843408.1 | XP_021699100.1 | 13 - 14 | 3559 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
