Assay Details
Gene Symbol: LOC5577895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842939.1 | XP_021698631.1 | 10 - 11 | 3241 | 60 | |
XM_021842942.1 | XP_021698634.1 | 10 - 11 | 3241 | 60 | ||
XM_021842944.1 | XP_021698636.1 | 10 - 11 | 3241 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
