Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661298.2 | XP_001661348.1 | 14 - 15 | 6486 | 105 | |
XM_021838871.1 | XP_021694563.1 | 15 - 16 | 6493 | 105 | ||
XM_021838872.1 | XP_021694564.1 | 14 - 15 | 6485 | 105 | ||
XM_021838873.1 | XP_021694565.1 | 15 - 16 | 6495 | 105 | ||
XM_021838874.1 | XP_021694566.1 | 15 - 16 | 6448 | 105 | ||
XM_021838875.1 | XP_021694567.1 | 15 - 16 | 6445 | 105 | ||
XM_021838876.1 | XP_021694568.1 | 14 - 15 | 4260 | 105 | ||
XM_021838877.1 | XP_021694569.1 | 14 - 15 | 4663 | 105 | ||
XM_021838878.1 | XP_021694570.1 | 14 - 15 | 4251 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
