Assay Details
Gene Symbol: LOC5574316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661298.2 | XP_001661348.1 | 21 - 22 | 7976 | 97 | |
XM_021838871.1 | XP_021694563.1 | 22 - 23 | 7995 | 97 | ||
XM_021838872.1 | XP_021694564.1 | 21 - 22 | 7987 | 97 | ||
XM_021838873.1 | XP_021694565.1 | 22 - 23 | 7985 | 97 | ||
XM_021838874.1 | XP_021694566.1 | 22 - 23 | 7950 | 97 | ||
XM_021838875.1 | XP_021694567.1 | 22 - 23 | 7947 | 97 | ||
XM_021838876.1 | XP_021694568.1 | 21 - 22 | 5762 | 97 | ||
XM_021838877.1 | XP_021694569.1 | 21 - 22 | 6165 | 97 | ||
XM_021838878.1 | XP_021694570.1 | 21 - 22 | 5753 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
