Assay Details
Gene Symbol: LOC5572902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847048.1 | XP_021702740.1 | 12 - 13 | 3519 | 60 | |
XM_021847049.1 | XP_021702741.1 | 12 - 13 | 3560 | 60 | ||
XM_021847050.1 | XP_021702742.1 | 12 - 13 | 3344 | 60 | ||
XM_021847051.1 | XP_021702743.1 | 12 - 13 | 3349 | 60 | ||
XM_021847052.1 | XP_021702744.1 | 12 - 13 | 3514 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
