Assay Details
Gene Symbol: LOC5572902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847048.1 | XP_021702740.1 | 14 - 15 | 3899 | 72 | |
XM_021847049.1 | XP_021702741.1 | 14 - 15 | 3940 | 72 | ||
XM_021847050.1 | XP_021702742.1 | 14 - 15 | 3724 | 72 | ||
XM_021847051.1 | XP_021702743.1 | 14 - 15 | 3729 | 72 | ||
XM_021847052.1 | XP_021702744.1 | 14 - 15 | 3894 | 72 |
