Assay Details
Gene Symbol: LOC5572902 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021847048.1 | XP_021702740.1 | 17 - 18 | 5335 | 70 | |
XM_021847049.1 | XP_021702741.1 | 17 - 18 | 5376 | 70 | ||
XM_021847050.1 | XP_021702742.1 | 17 - 18 | 5160 | 70 | ||
XM_021847051.1 | XP_021702743.1 | 17 - 18 | 5165 | 70 | ||
XM_021847052.1 | XP_021702744.1 | 17 - 18 | 5312 | 70 |

More Information
Set Membership: |
![]() ![]() |
