Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 11 - 12 | 2568 | 98 | |
XM_021844450.1 | XP_021700142.1 | 8 - 9 | 1917 | 98 |
