Assay Details
Gene Symbol: LOC5576280 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021844448.1 | XP_021700140.1 | 9 - 10 | 2049 | 108 | |
XM_021844449.1 | XP_021700141.1 | 8 - 9 | 2078 | 108 | ||
XM_021844450.1 | XP_021700142.1 | 6 - 7 | 1398 | 108 | ||
XM_021844451.1 | XP_021700143.1 | 8 - 9 | 2077 | 108 | ||
XM_021844452.1 | XP_021700144.1 | 8 - 9 | 2078 | 108 |

More Information
Set Membership: |
![]() ![]() |
