Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 1 - 2 | 978 | 121 | |
XM_021837702.1 | XP_021693394.1 | 1 - 2 | 978 | 121 | ||
XM_021837703.1 | XP_021693395.1 | 2 - 3 | 851 | 121 | ||
XM_021837704.1 | XP_021693396.1 | 2 - 3 | 842 | 121 | ||
XM_021837705.1 | XP_021693397.1 | 2 - 3 | 829 | 121 | ||
XM_021837706.1 | XP_021693398.1 | 1 - 2 | 974 | 121 | ||
XM_021837708.1 | XP_021693400.1 | 1 - 2 | 972 | 121 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021837701.1, XM_021837702.1, XM_021837703.1, XM_021837704.1, XM_021837705.1, XM_021837706.1, XM_021837708.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
