Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 6 - 7 | 2886 | 58 | |
XM_021837702.1 | XP_021693394.1 | 6 - 7 | 2802 | 58 | ||
XM_021837703.1 | XP_021693395.1 | 7 - 8 | 2758 | 58 | ||
XM_021837704.1 | XP_021693396.1 | 7 - 8 | 2749 | 58 | ||
XM_021837705.1 | XP_021693397.1 | 7 - 8 | 2736 | 58 | ||
XM_021837706.1 | XP_021693398.1 | 6 - 7 | 2881 | 58 | ||
XM_021837707.1 | XP_021693399.1 | 4 - 5 | 1190 | 58 | ||
XM_021837708.1 | XP_021693400.1 | 5 - 6 | 2223 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
