Assay Details
Gene Symbol: LOC23687451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837701.1 | XP_021693393.1 | 9 - 10 | 3290 | 75 | |
XM_021837702.1 | XP_021693394.1 | 9 - 10 | 3206 | 75 | ||
XM_021837703.1 | XP_021693395.1 | 10 - 11 | 3162 | 75 | ||
XM_021837704.1 | XP_021693396.1 | 10 - 11 | 3153 | 75 | ||
XM_021837705.1 | XP_021693397.1 | 10 - 11 | 3140 | 75 | ||
XM_021837706.1 | XP_021693398.1 | 8 - 9 | 3243 | 75 | ||
XM_021837707.1 | XP_021693399.1 | 7 - 8 | 1594 | 75 | ||
XM_021837708.1 | XP_021693400.1 | 8 - 9 | 2627 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
