Assay Details
Gene Symbol: LOC5569782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658670.2 | XP_001658720.2 | 1 - 2 | 884 | 62 | |
XM_021855723.1 | XP_021711415.1 | 2 - 3 | 757 | 62 | ||
XM_021855724.1 | XP_021711416.1 | 2 - 3 | 810 | 62 | ||
XM_021855725.1 | XP_021711417.1 | 2 - 3 | 862 | 62 | ||
XM_021855726.1 | XP_021711418.1 | 2 - 3 | 830 | 62 |

More Information
Set Membership: |
![]() ![]() |
