Assay Details
Gene Symbol: LOC5569782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658670.2 | XP_001658720.2 | 2 - 3 | 1041 | 67 | |
XM_021855721.1 | XP_021711413.1 | 2 - 3 | 1273 | 67 | ||
XM_021855722.1 | XP_021711414.1 | 3 - 4 | 2132 | 67 | ||
XM_021855723.1 | XP_021711415.1 | 3 - 4 | 914 | 67 | ||
XM_021855724.1 | XP_021711416.1 | 3 - 4 | 967 | 67 | ||
XM_021855725.1 | XP_021711417.1 | 3 - 4 | 1019 | 67 | ||
XM_021855726.1 | XP_021711418.1 | 3 - 4 | 987 | 67 | ||
XM_021855727.1 | XP_021711419.1 | 3 - 4 | 2280 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
