Assay Details
Gene Symbol: LOC5579338 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848420.1 | XP_021704112.1 | 7 - 8 | 1516 | 52 | |
XM_021848421.1 | XP_021704113.1 | 7 - 8 | 1450 | 52 | ||
XM_021848422.1 | XP_021704114.1 | 6 - 7 | 1495 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
