Assay Details
Gene Symbol: LOC5563715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837535.1 | XP_021693227.1 | 8 - 9 | 3748 | 67 | |
XM_021837536.1 | XP_021693228.1 | 7 - 8 | 3008 | 67 | ||
XM_021837537.1 | XP_021693229.1 | 8 - 9 | 3750 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
