Assay Details
Gene Symbol: LOC5574192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851655.1 | XP_021707347.1 | 11 - 12 | 15091 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851655.1 | XP_021707347.1 | 11 - 12 | 15091 | 65 |
Set Membership: |
![]() ![]() ![]() |