Assay Details
Gene Symbol: LOC5580104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848697.1 | XP_021704389.1 | 5 - 6 | 6285 | 63 | |
XM_021848704.1 | XP_021704396.1 | 5 - 6 | 6291 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
