Assay Details
Gene Symbol: LOC5576538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839109.1 | XP_021694801.1 | 2 - 3 | 1724 | 56 | |
XM_021839110.1 | XP_021694802.1 | 3 - 4 | 1469 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
