Assay Details
Gene Symbol: LOC5576538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839109.1 | XP_021694801.1 | 10 - 11 | 7477 | 93 | |
XM_021839110.1 | XP_021694802.1 | 11 - 12 | 7222 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
