Assay Details
Gene Symbol: LOC5576538 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839109.1 | XP_021694801.1 | 5 - 6 | 2873 | 63 | |
XM_021839110.1 | XP_021694802.1 | 6 - 7 | 2618 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
